Assay Details
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128431.4 | XP_003128479.1 | 561 | 114 | ||
| XM_005666100.3 | XP_005666157.1 | 424 | 114 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128431.4 | XP_003128479.1 | 561 | 114 | ||
| XM_005666100.3 | XP_005666157.1 | 424 | 114 | |||
Set Membership: |
|