Assay Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121465.4 | XP_003121513.1 | 4494 | 78 | ||
XM_013992893.2 | XP_013848347.1 | 4479 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
