Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073955.1 | XP_020929614.1 | 1152 | 74 | ||
| XM_021073956.1 | XP_020929615.1 | 1002 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073955.1 | XP_020929614.1 | 1152 | 74 | ||
| XM_021073956.1 | XP_020929615.1 | 1002 | 74 | |||
Set Membership: |
|