Assay Details
| Gene Symbol: MAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101277.1 | XP_020956936.1 | 2961 | 148 | ||
| XM_021101278.1 | XP_020956937.1 | 2136 | 148 | |||
| XM_021101279.1 | XP_020956938.1 | 2946 | 148 | |||
| XM_021101280.1 | XP_020956939.1 | 1645 | 148 | |||
More Information
Set Membership: |
|

MAP9
Genomic Map