Assay Details
| Gene Symbol: RAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661348.3 | XP_005661405.1 | 135 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005661348.3 transcripts.
Set Membership: |
|

RAX2
Genomic Map