Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123174.5 | XP_003123222.1 | 381 | 74 | ||
| XM_003123175.3 | XP_003123223.1 | 401 | 74 | |||
| XM_003123176.3 | XP_003123224.2 | 374 | 74 | |||
More Information
Set Membership: |
|

TMEM205
Genomic Map