Assay Details
| Gene Symbol: MECP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080802.1 | XP_020936461.1 | 83 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021080802.1 transcripts.
Set Membership: |
|

MECP2
Genomic Map