Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928637.4 | XP_001928672.1 | 275 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001928637.4 transcripts.
Set Membership: |
|

CHST3
Genomic Map