Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928637.4 | XP_001928672.1 | 487 | 104 | ||
| XM_005671049.3 | XP_005671106.1 | 448 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928637.4 | XP_001928672.1 | 487 | 104 | ||
| XM_005671049.3 | XP_005671106.1 | 448 | 104 | |||
Set Membership: |
|