Assay Details
| Gene Symbol: PURA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661730.3 | XP_005661787.1 | 347 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005661730.3 transcripts.
Set Membership: |
|

PURA
Genomic Map