Assay Details
| Gene Symbol: LOC100522551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664338.3 | XP_005664395.1 | 163 | 105 | ||
| XM_013998170.2 | XP_013853624.1 | 164 | 105 | |||
| XM_013998171.2 | XP_013853625.1 | 165 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005664338.3, XM_013998170.2, XM_013998171.2 transcripts.
Set Membership: |
|

LOC100522551
Genomic Map