Assay Details
| Gene Symbol: MAP3K7CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013982556.2 | XP_013838010.1 | 568 | 63 | ||
| XM_021070880.1 | XP_020926539.1 | 622 | 63 | |||
| XM_021070881.1 | XP_020926540.1 | 362 | 63 | |||
More Information
Set Membership: |
|

MAP3K7CL
Genomic Map