Assay Details
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093809.1 | XP_020949468.1 | 834 | 71 | ||
| XM_021093810.1 | XP_020949469.1 | 825 | 71 | |||
| XM_021093811.1 | XP_020949470.1 | 841 | 71 | |||
| XM_021093812.1 | XP_020949471.1 | 825 | 71 | |||

MTSS2
Genomic Map