Assay Details
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093810.1 | XP_020949469.1 | 552 | 76 | ||
| XM_021093812.1 | XP_020949471.1 | 552 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093810.1 | XP_020949469.1 | 552 | 76 | ||
| XM_021093812.1 | XP_020949471.1 | 552 | 76 | |||