Assay Details
| Gene Symbol: CCNYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672130.3 | XP_005672187.1 | 614 | 70 | ||
| XM_005672131.3 | XP_005672188.1 | 645 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCNYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672130.3 | XP_005672187.1 | 614 | 70 | ||
| XM_005672131.3 | XP_005672188.1 | 645 | 70 | |||
Set Membership: |
|