Assay Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131497.3 | XP_003131545.1 | 510 | 71 | ||
| XM_013979665.2 | XP_013835119.1 | 524 | 71 | |||
| XM_013979681.2 | XP_013835135.1 | 510 | 71 | |||
More Information
Set Membership: |
|

GSDMA
Genomic Map