Assay Details
| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078783.1 | XP_020934442.1 | 168 | 101 | ||
| XM_021078784.1 | XP_020934443.1 | 218 | 101 | |||
| XM_021078786.1 | XP_020934445.1 | 322 | 101 | |||
| XM_021078787.1 | XP_020934446.1 | 1601 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021078783.1, XM_021078784.1, XM_021078786.1, XM_021078787.1 transcripts.
Set Membership: |
|

ZNF775
Genomic Map