Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928633.6 | XP_001928668.1 | 543 | 120 | ||
| XM_005662871.3 | XP_005662928.1 | 446 | 120 | |||
| XM_013996502.2 | XP_013851956.1 | 323 | 120 | |||
| XM_013996503.2 | XP_013851957.1 | 536 | 120 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001928633.6, XM_005662871.3, XM_013996502.2, XM_013996503.2 transcripts.
Set Membership: |
|

ZHX1
Genomic Map