Assay Details
| Gene Symbol: GPR132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656481.3 | XP_005656538.1 | 572 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005656481.3 transcripts.
Set Membership: |
|

GPR132
Genomic Map