Assay Details
| Gene Symbol: C7H14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099247.1 | XP_020954906.1 | 1697 | 109 | ||
| XM_021099248.1 | XP_020954907.1 | 3713 | 109 | |||
More Information
Set Membership: |
|

C7H14orf93
Genomic Map