Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122506.5 | XP_003122554.1 | 204 | 110 | ||
| XM_021082815.1 | XP_020938474.1 | 213 | 110 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003122506.5, XM_021082815.1 transcripts.
Set Membership: |
|

GAL3ST3
Genomic Map