Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122506.5 | XP_003122554.1 | 448 | 81 | ||
| XM_005660669.3 | XP_005660726.1 | 374 | 81 | |||
| XM_021082814.1 | XP_020938473.1 | 1219 | 81 | |||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map