Assay Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481897.4 | XP_003481945.2 | 642 | 95 | ||
| XM_005664753.3 | XP_005664810.1 | 364 | 95 | |||
| XM_005664754.3 | XP_005664811.1 | 345 | 95 | |||
| XM_013998675.2 | XP_013854129.1 | 297 | 95 | |||
| XM_021094762.1 | XP_020950421.1 | 1475 | 95 | |||
| XM_021094763.1 | XP_020950422.1 | 1419 | 95 | |||
| XM_021094764.1 | XP_020950423.1 | 446 | 95 | |||
More Information
Set Membership: |
|

IL4I1
Genomic Map