Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069798.1 | XP_020925457.1 | 574 | 95 | ||
| XM_021069799.1 | XP_020925458.1 | 574 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069798.1 | XP_020925457.1 | 574 | 95 | ||
| XM_021069799.1 | XP_020925458.1 | 574 | 95 | |||
Set Membership: |
|