Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928669.7 | XP_001928704.1 | 253 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001928669.7 transcripts.
Set Membership: |
|

SEPTIN3
Genomic Map