Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928669.7 | XP_001928704.1 | 663 | 117 | ||
| XM_021091436.1 | XP_020947095.1 | 1077 | 117 | |||
| XM_021091437.1 | XP_020947096.1 | 1077 | 117 | |||
| XM_021091438.1 | XP_020947097.1 | 1077 | 117 | |||
| GenBank mRNA | KU358535.1 | - | 4 - 5 | 437 | 117 | |
More Information
Set Membership: |
|

SEPTIN3
Genomic Map