Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928669.7 | XP_001928704.1 | 1112 | 70 | ||
| XM_021091436.1 | XP_020947095.1 | 1526 | 70 | |||
| XM_021091437.1 | XP_020947096.1 | 1523 | 70 | |||
| XM_021091438.1 | XP_020947097.1 | 1526 | 70 | |||
| GenBank mRNA | KU358535.1 | - | 8 - 9 | 886 | 70 | |
More Information
Set Membership: |
|

SEPTIN3
Genomic Map