Assay Details
| Gene Symbol: DUSP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092338.1 | XP_020947997.1 | 1253 | 72 | ||
| XM_021092339.1 | XP_020947998.1 | 1317 | 72 | |||
| XM_021092340.1 | XP_020947999.1 | 1277 | 72 | |||
| XM_021092341.1 | XP_020948000.1 | 1186 | 72 | |||
More Information
Set Membership: |
|

DUSP16
Genomic Map