Assay Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071034.1 | XP_020926693.1 | 958 | 79 | ||
| XM_021071035.1 | XP_020926694.1 | 935 | 79 | |||
| XM_021071036.1 | XP_020926695.1 | 1028 | 79 | |||
| XM_021071037.1 | XP_020926696.1 | 1040 | 79 | |||
More Information
Set Membership: |
|

RSPH1
Genomic Map