Assay Details
| Gene Symbol: GPR176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097914.1 | XP_020953573.1 | 542 | 75 | ||
| XM_021097925.1 | XP_020953584.1 | 542 | 75 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021097914.1, XM_021097925.1 transcripts.
Set Membership: |
|

GPR176
Genomic Map