Assay Details
| Gene Symbol: LOC100523684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062993.1 | XP_020918652.1 | 462 | 63 | ||
| XM_021062994.1 | XP_020918653.1 | 386 | 63 | |||
| XM_021062995.1 | XP_020918654.1 | 593 | 63 | |||
| XM_021062996.1 | XP_020918655.1 | 516 | 63 | |||
| XM_021062997.1 | XP_020918656.1 | 440 | 63 | |||
| XM_021062998.1 | XP_020918657.1 | 267 | 63 | |||
| XM_021062999.1 | XP_020918658.1 | 1813 | 63 | |||
| XM_021063000.1 | XP_020918659.1 | 379 | 63 | |||
| XM_021063001.1 | XP_020918660.1 | 1867 | 63 | |||
| XM_021063002.1 | XP_020918661.1 | 239 | 63 | |||
| XM_021063007.1 | XP_020918666.1 | 441 | 63 | |||
| XM_021063008.1 | XP_020918667.1 | 365 | 63 | |||
More Information
Set Membership: |
|

LOC100523684
Genomic Map