Assay Details
| Gene Symbol: NDUFAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925041.6 | XP_001925076.4 | 3807 | 100 | ||
| XM_021089081.1 | XP_020944740.1 | 3728 | 100 | |||
More Information
Set Membership: |
|

NDUFAF6
Genomic Map