Assay Details
| Gene Symbol: LOC110255897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066302.1 | XP_020921961.1 | 736 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110255897| Gene Symbol: LOC110255897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066302.1 | XP_020921961.1 | 736 | 92 | ||