Assay Details
| Gene Symbol: CENPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133338.5 | XP_003133386.3 | 428 | 76 | ||
| XM_005671710.3 | XP_005671767.2 | 159 | 76 | |||
| XM_013984043.2 | XP_013839497.2 | 430 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005671710.3 transcripts.
Set Membership: |
|

CENPU
Genomic Map