Assay Details
| Gene Symbol: CRLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083475.1 | XP_020939134.1 | 539 | 64 | ||
| XM_021083476.1 | XP_020939135.1 | 539 | 64 | |||
| XM_021083477.1 | XP_020939136.1 | 539 | 64 | |||
More Information
Set Membership: |
|

CRLF1
Genomic Map