Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653856.3 | XP_005653913.2 | 1610 | 85 | ||
| GenBank mRNA | KU358542.1 | - | 8 - 9 | 1483 | 85 | |
More Information
Set Membership: |
|

SEPTIN9
Genomic Map