Assay Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086276.1 | XP_020941935.1 | 1450 | 60 | ||
| GenBank mRNA | KU705639.1 | - | 10 - 11 | 1294 | 60 | |
More Information
Set Membership: |
|

LRWD1
Genomic Map