Assay Details
| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481278.4 | XP_003481326.1 | 260 | 86 | ||
| XM_021087775.1 | XP_020943434.1 | 282 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC27| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481278.4 | XP_003481326.1 | 260 | 86 | ||
| XM_021087775.1 | XP_020943434.1 | 282 | 86 | |||
Set Membership: |
|