Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132654.4 | XP_003132702.1 | 2049 | 109 | ||
| XM_013982409.2 | XP_013837863.1 | 2144 | 109 | |||
| XM_021070320.1 | XP_020925979.1 | 2049 | 109 | |||
| XM_021070321.1 | XP_020925980.1 | 2049 | 109 | |||
| XM_021070322.1 | XP_020925981.1 | 1950 | 109 | |||
More Information
Set Membership: |
|

CFAP91
Genomic Map