Assay Details
| Gene Symbol: PIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093742.1 | XP_020949401.1 | 770 | 64 | ||
| XM_021093751.1 | XP_020949410.1 | 770 | 64 | |||
| XM_021093760.1 | XP_020949419.1 | 810 | 64 | |||
| XM_021093763.1 | XP_020949422.1 | 770 | 64 | |||
| XM_021093764.1 | XP_020949423.1 | 770 | 64 | |||
| XM_021093766.1 | XP_020949425.1 | 809 | 64 | |||
| XM_021093769.1 | XP_020949428.1 | 1332 | 64 | |||
More Information
Set Membership: |
|

PIF1
Genomic Map