Assay Details
| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657884.3 | XP_005657941.1 | 293 | 96 | ||
| XM_005657885.3 | XP_005657942.1 | 320 | 96 | |||
| XM_013990972.2 | XP_013846426.1 | 293 | 96 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005657884.3, XM_005657885.3, XM_013990972.2 transcripts.
Set Membership: |
|

BHLHB9
Genomic Map