Assay Details
| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135260.5 | XP_003135308.1 | 398 | 119 | ||
| XM_005657883.3 | XP_005657940.1 | 475 | 119 | |||
| XM_005657884.3 | XP_005657941.1 | 500 | 119 | |||
| XM_005657885.3 | XP_005657942.1 | 527 | 119 | |||
| XM_005657888.3 | XP_005657945.1 | 340 | 119 | |||
| XM_005657890.3 | XP_005657947.1 | 352 | 119 | |||
| XM_013990972.2 | XP_013846426.1 | 484 | 119 | |||
| XM_021079936.1 | XP_020935595.1 | 454 | 119 | |||
| XM_021079937.1 | XP_020935596.1 | 340 | 119 | |||
| XM_021079938.1 | XP_020935597.1 | 341 | 119 | |||
| XM_021079939.1 | XP_020935598.1 | 396 | 119 | |||
| XM_021079940.1 | XP_020935599.1 | 324 | 119 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135260.5, XM_005657883.3, XM_005657884.3, XM_005657885.3, XM_005657888.3, XM_005657890.3, XM_013990972.2, XM_021079936.1, XM_021079937.1, XM_021079938.1, XM_021079939.1, XM_021079940.1 transcripts.
Set Membership: |
|

BHLHB9
Genomic Map