Assay Details
| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135260.5 | XP_003135308.1 | 473 | 79 | ||
| XM_005657883.3 | XP_005657940.1 | 550 | 79 | |||
| XM_005657884.3 | XP_005657941.1 | 575 | 79 | |||
| XM_005657885.3 | XP_005657942.1 | 602 | 79 | |||
| XM_005657888.3 | XP_005657945.1 | 415 | 79 | |||
| XM_005657890.3 | XP_005657947.1 | 427 | 79 | |||
| XM_013990972.2 | XP_013846426.1 | 559 | 79 | |||
| XM_021079936.1 | XP_020935595.1 | 529 | 79 | |||
| XM_021079937.1 | XP_020935596.1 | 415 | 79 | |||
| XM_021079938.1 | XP_020935597.1 | 416 | 79 | |||
| XM_021079939.1 | XP_020935598.1 | 471 | 79 | |||
| XM_021079940.1 | XP_020935599.1 | 399 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135260.5, XM_005657883.3, XM_005657884.3, XM_005657885.3, XM_005657888.3, XM_005657890.3, XM_013990972.2, XM_021079936.1, XM_021079937.1, XM_021079938.1, XM_021079939.1, XM_021079940.1 transcripts.
Set Membership: |
|

BHLHB9
Genomic Map