Assay Details
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126907.4 | XP_003126955.1 | 1078 | 123 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
