Assay Details
| Gene Symbol: C2H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661243.3 | XP_005661300.1 | 359 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005661243.3 transcripts.
Set Membership: |
|

C2H19orf57
Genomic Map