Assay Details
Gene Symbol: ZNF581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005664852.3 | XP_005664909.1 | 4911 | 103 | ||
XM_021095059.1 | XP_020950718.1 | 601 | 103 | |||
XM_021095061.1 | XP_020950720.1 | 412 | 103 | |||
XM_021095062.1 | XP_020950721.1 | 353 | 103 | |||
XM_021095063.1 | XP_020950722.1 | 345 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005664852.3, XM_021095059.1, XM_021095061.1, XM_021095062.1, XM_021095063.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
