Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092202.1 | XP_020947861.1 | 1666 | 131 | ||
| XM_021092203.1 | XP_020947862.1 | 1931 | 131 | |||
| XM_021092204.1 | XP_020947863.1 | 1662 | 131 | |||
| XM_021092206.1 | XP_020947865.1 | 1640 | 131 | |||
More Information
Set Membership: |
|

IRAG2
Genomic Map