Assay Details
| Gene Symbol: LTB4R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927733.6 | XP_001927768.3 | 771 | 105 | ||
| XM_021099203.1 | XP_020954862.1 | 700 | 105 | |||
| XM_021099204.1 | XP_020954863.1 | 619 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001927733.6, XM_021099203.1, XM_021099204.1 transcripts.
Set Membership: |
|

LTB4R2
Genomic Map