Assay Details
| Gene Symbol: TBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088013.1 | XP_020943672.1 | 1709 | 82 | ||
| XM_021088014.1 | XP_020943673.1 | 1663 | 82 | |||
| XM_021088015.1 | XP_020943674.1 | 1496 | 82 | |||
| XM_021088016.1 | XP_020943675.1 | 1190 | 82 | |||
| XM_021088017.1 | XP_020943676.1 | 1205 | 82 | |||
| XM_021088018.1 | XP_020943677.1 | 1190 | 82 | |||
More Information
Set Membership: |
|

TBX6
Genomic Map