Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013990082.2 | XP_013845536.2 | 100 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013990082.2 transcripts.
Set Membership: |
|

ZKSCAN7
Genomic Map